Anti-AGR2 Mouse Monoclonal Antibody [clone: 1C9]

Supplier: Abnova
H00010551-M05
10510-520EA 335 USD
10510-520
Anti-AGR2 Mouse Monoclonal Antibody [clone: 1C9]
Antibodies
Type: Primary
Antigen: AGR2
Clonality: Monoclonal
Clone: 1C9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR