Anti-PROCR Mouse Monoclonal Antibody [clone: M2]

Supplier: Abnova
H00010544-M03
10598-212EA 335 USD
10598-212
Anti-PROCR Mouse Monoclonal Antibody [clone: M2]
Antibodies
Type: Primary
Antigen: PROCR
Clonality: Monoclonal
Clone: M2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR