Anti-PROCR Mouse Monoclonal Antibody [clone: M1-5A10]

Supplier: Abnova
H00010544-M01
10598-210EA 335 USD
10598-210
Anti-PROCR Mouse Monoclonal Antibody [clone: M1-5A10]
Antibodies
Type: Primary
Antigen: PROCR
Clonality: Monoclonal
Clone: M1-5A10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR