Anti-LAMTOR5 Mouse Monoclonal Antibody [clone: 3D3]

Supplier: Abnova
H00010542-M02A
10624-462EA 335 USD
10624-462
Anti-LAMTOR5 Mouse Monoclonal Antibody [clone: 3D3]
Antibodies
Type: Primary
Antigen: LAMTOR5
Clonality: Monoclonal
Clone: 3D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR