Anti-UBD Mouse Monoclonal Antibody [clone: 7D8]

Supplier: Abnova
H00010537-M01
10573-286EA 335 USD
10573-286
Anti-UBD Mouse Monoclonal Antibody [clone: 7D8]
Antibodies
Type: Primary
Antigen: UBD
Clonality: Monoclonal
Clone: 7D8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR