Anti-PITRM1 Mouse Monoclonal Antibody [clone: 1H3]

Supplier: Abnova
H00010531-M03
10597-062EA 335 USD
10597-062
Anti-PITRM1 Mouse Monoclonal Antibody [clone: 1H3]
Antibodies
Type: Primary
Antigen: PITRM1
Clonality: Monoclonal
Clone: 1H3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR