Anti-HYOU1 Mouse Monoclonal Antibody [clone: 6F7]

Supplier: Abnova
H00010525-M01
10589-622EA 335 USD
10589-622
Anti-HYOU1 Mouse Monoclonal Antibody [clone: 6F7]
Antibodies
Type: Primary
Antigen: HYOU1
Clonality: Monoclonal
Clone: 6F7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR