Anti-DEAF1 Mouse Monoclonal Antibody [clone: 2A2]

Supplier: Abnova
H00010522-M07
10583-776EA 335 USD
10583-776
Anti-DEAF1 Mouse Monoclonal Antibody [clone: 2A2]
Antibodies
Type: Primary
Antigen: DEAF1
Clonality: Monoclonal
Clone: 2A2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR