Anti-SEMA4D Mouse Monoclonal Antibody [clone: 3B4]

Supplier: Abnova
H00010507-M01
10600-902EA 335 USD
10600-902
Anti-SEMA4D Mouse Monoclonal Antibody [clone: 3B4]
Antibodies
Type: Primary
Antigen: SEMA4D
Clonality: Monoclonal
Clone: 3B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR