Anti-CARM1 Mouse Polyclonal Antibody

Supplier: Abnova
H00010498-A01
10552-538EA 243 USD
10552-538
Anti-CARM1 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CARM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR