Anti-VAT1 Mouse Monoclonal Antibody [clone: 3E9]

Supplier: Abnova
H00010493-M07
10604-832EA 335 USD
10604-832
Anti-VAT1 Mouse Monoclonal Antibody [clone: 3E9]
Antibodies
Type: Primary
Antigen: VAT1
Clonality: Monoclonal
Clone: 3E9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR