Anti-FST Mouse Monoclonal Antibody [clone: 4B11]

Supplier: Abnova
H00010468-M10
10586-784EA 335 USD
10586-784
Anti-FST Mouse Monoclonal Antibody [clone: 4B11]
Antibodies
Type: Primary
Antigen: FST (follistatin)
Clonality: Monoclonal
Clone: 4B11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR