Anti-PIBF1 Mouse Monoclonal Antibody [clone: 7B7]

Supplier: Abnova
H00010464-M02
10578-234EA 335 USD
10578-234
Anti-PIBF1 Mouse Monoclonal Antibody [clone: 7B7]
Antibodies
Type: Primary
Antigen: PIBF1
Clonality: Monoclonal
Clone: 7B7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR