Anti-BAIAP2 Mouse Monoclonal Antibody [clone: 3F3-2D3]

Supplier: Abnova
H00010458-M01
10577-764EA 335 USD
10577-764
Anti-BAIAP2 Mouse Monoclonal Antibody [clone: 3F3-2D3]
Antibodies
Type: Primary
Antigen: BAIAP2
Clonality: Monoclonal
Clone: 3F3-2D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR