Anti-OLFM1 Mouse Monoclonal Antibody [clone: 2E9-1A2]

Supplier: Abnova
H00010439-M02
10595-672EA 335 USD
10595-672
Anti-OLFM1 Mouse Monoclonal Antibody [clone: 2E9-1A2]
Antibodies
Type: Primary
Antigen: OLFM1
Clonality: Monoclonal
Clone: 2E9-1A2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR