Anti-CDC42EP2 Mouse Monoclonal Antibody [clone: 2H7]

Supplier: Abnova
H00010435-M01
10581-654EA 335 USD
10581-654
Anti-CDC42EP2 Mouse Monoclonal Antibody [clone: 2H7]
Antibodies
Type: Primary
Antigen: CDC42EP2
Clonality: Monoclonal
Clone: 2H7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR