Anti-ARIH2 Mouse Monoclonal Antibody [clone: 1C3]

Supplier: Abnova
H00010425-M01A
10620-596EA 335 USD
10620-596
Anti-ARIH2 Mouse Monoclonal Antibody [clone: 1C3]
Antibodies
Type: Primary
Antigen: ARIH2
Clonality: Monoclonal
Clone: 1C3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR