Anti-TESK2 Mouse Monoclonal Antibody [clone: 5D1]

Supplier: Abnova
H00010420-M05
10603-576EA 335 USD
10603-576
Anti-TESK2 Mouse Monoclonal Antibody [clone: 5D1]
Antibodies
Type: Primary
Antigen: TESK2
Clonality: Monoclonal
Clone: 5D1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR