Anti-PRMT5 Mouse Monoclonal Antibody [clone: 4F2]

Supplier: Abnova
H00010419-M03
10601-020EA 335 USD
10601-020
Anti-PRMT5 Mouse Monoclonal Antibody [clone: 4F2]
Antibodies
Type: Primary
Antigen: PRMT5
Clonality: Monoclonal
Clone: 4F2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR