Anti-YAP1 Mouse Monoclonal Antibody [clone: 2F12]

Supplier: Abnova
H00010413-M01
10605-974EA 335 USD
10605-974
Anti-YAP1 Mouse Monoclonal Antibody [clone: 2F12]
Antibodies
Type: Primary
Antigen: YAP1
Clonality: Monoclonal
Clone: 2F12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR