Anti-RAPGEF3 Mouse Monoclonal Antibody [clone: 2E5]

Supplier: Abnova
H00010411-M01
10599-162EA 335 USD
10599-162
Anti-RAPGEF3 Mouse Monoclonal Antibody [clone: 2E5]
Antibodies
Type: Primary
Antigen: RAPGEF3
Clonality: Monoclonal
Clone: 200000
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR