Anti-CORO2B Mouse Polyclonal Antibody

Supplier: Abnova
H00010391-B01P
10553-966EA 335 USD
10553-966
Anti-CORO2B Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CORO2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR