Anti-SCML2 Mouse Monoclonal Antibody [clone: 1C8]

Supplier: Abnova
H00010389-M01
10600-738EA 335 USD
10600-738
Anti-SCML2 Mouse Monoclonal Antibody [clone: 1C8]
Antibodies
Type: Primary
Antigen: SCML2
Clonality: Monoclonal
Clone: 1C8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR