Anti-TUBB2A Mouse Monoclonal Antibody [clone: 1F9]

Supplier: Abnova
H00010383-M01
10605-056EA 335 USD
10605-056
Anti-TUBB2A Mouse Monoclonal Antibody [clone: 1F9]
Antibodies
Type: Primary
Antigen: TUBB2A
Clonality: Monoclonal
Clone: 1F9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR