Anti-BPNT1 Mouse Monoclonal Antibody [clone: 2E1]

Supplier: Abnova
H00010380-M01
10578-054EA 335 USD
10578-054
Anti-BPNT1 Mouse Monoclonal Antibody [clone: 2E1]
Antibodies
Type: Primary
Antigen: BPNT1
Clonality: Monoclonal
Clone: 2E1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR