Anti-IRF9 Mouse Monoclonal Antibody [clone: 1D11]

Supplier: Abnova
H00010379-M02
10590-710EA 335 USD
10590-710
Anti-IRF9 Mouse Monoclonal Antibody [clone: 1D11]
Antibodies
Type: Primary
Antigen: IRF9
Clonality: Monoclonal
Clone: 1D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR