Anti-IRF9 Rabbit Polyclonal Antibody

Supplier: Abnova
H00010379-D01
10590-712EA 376 USD
10590-712
Anti-IRF9 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: IRF9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR