Anti-CACNG3 Mouse Monoclonal Antibody [clone: 30000]

Supplier: Abnova
H00010368-M01
10578-476EA 335 USD
10578-476
Anti-CACNG3 Mouse Monoclonal Antibody [clone: 30000]
Antibodies
Type: Primary
Antigen: CACNG3
Clonality: Monoclonal
Clone: 30000
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR