Anti-KLF2 Mouse Polyclonal Antibody

Supplier: Abnova
H00010365-A01
10562-492EA 243 USD
10562-492
Anti-KLF2 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: KLF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR