Anti-NPM3 Mouse Monoclonal Antibody [clone: 3F2]

Supplier: Abnova
H00010360-M02
10595-204EA 335 USD
10595-204
Anti-NPM3 Mouse Monoclonal Antibody [clone: 3F2]
Antibodies
Type: Primary
Antigen: NPM3
Clonality: Monoclonal
Clone: 3F2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR