Anti-TLR6 Mouse Monoclonal Antibody [clone: 4F4]

Supplier: Abnova
H00010333-M11
10604-112EA 335 USD
10604-112
Anti-TLR6 Mouse Monoclonal Antibody [clone: 4F4]
Antibodies
Type: Primary
Antigen: TLR6
Clonality: Monoclonal
Clone: 4F4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR