Anti-AKR1A1 Mouse Monoclonal Antibody [clone: 1A11-2A4]

Supplier: Abnova

H00010327-M01
10576-540EA 325 USD
10576-540
Anti-AKR1A1 Mouse Monoclonal Antibody [clone: 1A11-2A4]
Antibodies
Type: Primary
Antigen: AKR1A1
Clonality: Monoclonal
Clone: 1A11-2A4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR