Anti-AKR1A1 Mouse Polyclonal Antibody

Supplier: Abnova
H00010327-B01
10510-610EA 335 USD
10510-610
Anti-AKR1A1 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: AKR1A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR