Anti-KLHL41 Mouse Monoclonal Antibody [clone: 1H3]

Supplier: Abnova
H00010324-M01
10590-920EA 335 USD
10590-920
Anti-KLHL41 Mouse Monoclonal Antibody [clone: 1H3]
Antibodies
Type: Primary
Antigen: KLHL41
Clonality: Monoclonal
Clone: 1H3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR