Anti-PAK4 Mouse Monoclonal Antibody [clone: 3F10]

Supplier: Abnova

H00010298-M01
10595-948EA 325 USD
10595-948
Anti-PAK4 Mouse Monoclonal Antibody [clone: 3F10]
Antibodies
Type: Primary
Antigen: PAK4
Clonality: Monoclonal
Clone: 3F10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR