Anti-DNAJ Mouse Monoclonal Antibody [clone: 2A11-F2]

Supplier: Abnova
H00010294-M01
10584-172EA 335 USD
10584-172
Anti-DNAJ Mouse Monoclonal Antibody [clone: 2A11-F2]
Antibodies
Type: Primary
Antigen: DNAJ
Clonality: Monoclonal
Clone: 2A11-F2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR