Anti-LILRB2 Mouse Monoclonal Antibody [clone: 1D4]

Supplier: Abnova
H00010288-M01
10591-808EA 335 USD
10591-808
Anti-LILRB2 Mouse Monoclonal Antibody [clone: 1D4]
Antibodies
Type: Primary
Antigen: LILRB2
Clonality: Monoclonal
Clone: 1D4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR