Anti-SMNDC1 Mouse Polyclonal Antibody

Supplier: Abnova
H00010285-B01P
10571-668EA 335 USD
10571-668
Anti-SMNDC1 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SMNDC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR