Anti-STUB1 Mouse Monoclonal Antibody [clone: 2E12]

Supplier: Abnova
H00010273-M01
10602-808EA 335 USD
10602-808
Anti-STUB1 Mouse Monoclonal Antibody [clone: 2E12]
Antibodies
Type: Primary
Antigen: STUB1
Clonality: Monoclonal
Clone: 2E12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR