Anti-RAMP3 Mouse Monoclonal Antibody [clone: 1C11]

Supplier: Abnova

H00010268-M01
10599-134EA 325 USD
10599-134
Anti-RAMP3 Mouse Monoclonal Antibody [clone: 1C11]
Antibodies
Type: Primary
Antigen: RAMP3
Clonality: Monoclonal
Clone: 1C11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR