Anti-IRX5 Mouse Monoclonal Antibody [clone: 8A12]

Supplier: Abnova

H00010265-M06
10590-682EA 325 USD
10590-682
Anti-IRX5 Mouse Monoclonal Antibody [clone: 8A12]
Antibodies
Type: Primary
Antigen: IRX5
Clonality: Monoclonal
Clone: 8A12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR