Anti-ABCC4 Mouse Monoclonal Antibody [clone: 1B2]

Supplier: Abnova
H00010257-M03
10575-976EA 335 USD
10575-976
Anti-ABCC4 Mouse Monoclonal Antibody [clone: 1B2]
Antibodies
Type: Primary
Antigen: ABCC4
Clonality: Monoclonal
Clone: 1B2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR