Anti-SPRY2 Mouse Monoclonal Antibody [clone: 1E10]

Supplier: Abnova
H00010253-M01
10602-028EA 335 USD
10602-028
Anti-SPRY2 Mouse Monoclonal Antibody [clone: 1E10]
Antibodies
Type: Primary
Antigen: SPRY2
Clonality: Monoclonal
Clone: 1E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR