Anti-POP7 Mouse Monoclonal Antibody [clone: 3F12]

Supplier: Abnova
H00010248-M13
10597-512EA 335 USD
10597-512
Anti-POP7 Mouse Monoclonal Antibody [clone: 3F12]
Antibodies
Type: Primary
Antigen: POP7
Clonality: Monoclonal
Clone: 3F12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR