Anti-CALCOCO2 Mouse Monoclonal Antibody [clone: 3H6]

Supplier: Abnova
H00010241-M05
10594-524EA 335 USD
10594-524
Anti-CALCOCO2 Mouse Monoclonal Antibody [clone: 3H6]
Antibodies
Type: Primary
Antigen: CALCOCO2
Clonality: Monoclonal
Clone: 3H6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR