Anti-CD96 Rabbit Polyclonal Antibody

Supplier: Abnova
H00010225-D01
10581-552EA 376 USD
10581-552
Anti-CD96 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CD96
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR