Anti-TRIB1 Mouse Monoclonal Antibody [clone: 6E5]

Supplier: Abnova
H00010221-M02
10603-896EA 335 USD
10603-896
Anti-TRIB1 Mouse Monoclonal Antibody [clone: 6E5]
Antibodies
Type: Primary
Antigen: TRIB1
Clonality: Monoclonal
Clone: 600000
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR