Anti-OLIG2 Mouse Monoclonal Antibody [clone: 2B11]

Supplier: Abnova
H00010215-M06
10595-694EA 335 USD
10595-694
Anti-OLIG2 Mouse Monoclonal Antibody [clone: 2B11]
Antibodies
Type: Primary
Antigen: OLIG2
Clonality: Monoclonal
Clone: 2B11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR