Anti-TOPORS (topoisomerase I binding, arginine/serine-rich, E3 ubiquitin Protein ligase) Mouse Monoclonal Antibody [clone: 5G11]

Supplier: Abnova
H00010210-M01
10604-434EA 335 USD
10604-434
Anti-TOPORS (topoisomerase I binding, arginine/serine-rich, E3 ubiquitin Protein ligase) Mouse Monoclonal Antibody [clone: 5G11]
Antibodies
Type: Primary
Antigen: TOPORS (topoisomerase I binding, arginine/serine-rich, E3 ubiquitin Protein ligase)
Clonality: Monoclonal
Clone: 5G11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR