Anti-EIF1 Mouse Monoclonal Antibody [clone: 20]

Supplier: Abnova
H00010209-M01A
10625-270EA 335 USD
10625-270
Anti-EIF1 Mouse Monoclonal Antibody [clone: 20]
Antibodies
Type: Primary
Antigen: EIF1
Clonality: Monoclonal
Clone: 20
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR